Merck
  • Simultaneous determination of bendamustine and its active metabolite, gamma-hydroxy-bendamustine in human plasma and urine using HPLC-fluorescence detector: application to a pharmacokinetic study in Chinese cancer patients.
Simultaneous determination of bendamustine and its active metabolite, gamma-hydroxy-bendamustine in human plasma and urine using HPLC-fluorescence detector: application to a pharmacokinetic study in Chinese cancer patients.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-05-06)
Feifan Xie, Zeneng Cheng, Hang Cheng, Peng Yu
ABSTRACT
A simple, sensitive and cost-effective assay based on reversed phase high performance liquid chromatography (RP-HPLC) with isocratic mode for simultaneous determination of bendamustine (BM) and its active metabolite, gamma-hydroxy-bendamustine (γ-OH-BM) in human plasma and urine was developed and validated. Sample preparation involved protein precipitation by 10% perchloric acid-methanol solution. The peaks were recorded by using fluorescence detector (excitation wavelength 328 nm and emission wavelength 420 nm). The calibration curves were linear over concentration ranges of 8.192-10,000 ng mL(-1) and 5-1,000 ng mL(-1) for BM in human plasma and urine as well as 10-1,000 ng mL(-1) and 5-1,000 ng mL(-1) for γ-OH-BM in human plasma and urine, respectively. Intra- and inter-run precisions of BM and γ-OH-BM were less than 15% and the bias were within ± 15% for both plasma and urine. This validated method was successfully applied to a pharmacokinetic study enrolling 10 Chinese patients with indolent B-cell non-Hodgkin lymphoma and chronic lymphocytic leukemia administered a single intravenous infusion of 100 mg m(2) bendamustine hydrochloride.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Valeric acid, ≥99%
Sigma-Aldrich
Valeric acid, ≥99%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Valeric acid, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%