Merck
  • A victory for statins or a defeat for diet policies? Cholesterol falls in Poland in the past decade: A modeling study.
A victory for statins or a defeat for diet policies? Cholesterol falls in Poland in the past decade: A modeling study.
International journal of cardiology (2015-04-02)
Piotr Bandosz, Martin O'Flaherty, Marcin Rutkowski, Chris Kypridemos, Maria Guzman-Castillo, Duncan O S Gillespie, Bogdan Solnica, Michael J Pencina, Bogdan Wyrzykowski, Simon Capewell, Tomasz Zdrojewski
ABSTRACT
The present study is aimed to examine whether recent changes in population total cholesterol (TC) levels in Poland might be attributed to increased use statins. Two independent, nationally representative cross-sectional studies were conducted in adults aged 18-79 years in 2002 (n=2993, mean age 46.2 years) and 2011 (n=2413, mean age 45.8 years), including measurements of TC in venous blood samples. The mean change of TC between 2002 and 2011 was assessed. Then the expected therapeutic reduction in TC level in 2011 attributable to statins only was calculated based on already published statin effectiveness data. Uncertainty was quantified using probabilistic sensitivity analysis. Statin uptake in Poland rose to 11.2% in 2011 (95% Confidence Intervals (CI): 10% to 12.5%) and approximately 32% (95% CI: 28.4 to 36.0%) in subjects aged 60-79 years. Mean TC in Poland in 2002 was 5.35 mmol/l, and fell by 0.21 mmol/l (95% CI: 0.14 to 0.28) by 2011. This fall would have been only 0.03 mmol/l (95% CI: -0.04 to 0.10) for the total adult population and 0.06 mmol/l (95% CI: -0.09 to 0.22) in people aged 60-79 years if statins had not been used. Statin use thus apparently explained approximately 85% (95% CI: 49% to 120%) of the observed decrease. Between 2002 and 2011, statin medications apparently explained a large part of the observed fall in population cholesterol level, suggesting very little changes in population TC attributed to dietary changes.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material