Merck
  • A novel cation exchange polymer as a reversed-dispersive solid phase extraction sorbent for the rapid determination of rhodamine B residue in chili powder and chili oil.
A novel cation exchange polymer as a reversed-dispersive solid phase extraction sorbent for the rapid determination of rhodamine B residue in chili powder and chili oil.
Journal of chromatography. A (2014-12-17)
Dawei Chen, Yunfeng Zhao, Hong Miao, Yongning Wu
ABSTRACT
This paper presents a new analytical method for the determination of rhodamine B (RB) residue in chili powder and chili oil based on a novel reversed-dispersive solid phase extraction (r-dSPE) and ultra high performance liquid chromatography–high resolution mass spectrometry (UHPLC–HRMS). Chili powder and chili oil samples were first extracted with acetonitrile/water (1:1, v/v) and acetonitrile, respectively. Then, RB from the extract was adsorbed to the polymer cation exchange (PCX) sorbent with the characteristics of ion exchange and reversed-phase retention. Subsequently, the analyte in PCX sorbent was eluted with ammonium hydroxide/methanol (1:99, v/v) through a simple unit device equipped with 1 mL syringe and 0.22 μm nylon syringe filter. All of the samples were analyzed by UHPLC–HRMS/MS on a Waters Acquity BEH C18 column with 0.1% formic acid and 4 mM ammonium formate in water/acetonitrile as the mobile phase with gradient elution. The matrix effect, recovery, and repeatability, within laboratory reproducibility, and the LODs and LOQs of the r-dSPE cleanup method were investigated. The method showed a good linearity (R2 > 0.999) in the ranges of 0.01–1 μg/L and 1–100 μg/L for the analyte. The LODs of RB for chili powder and chili oil samples were 0.5 μg/kg. The average recoveries of RB from the samples spiked at four different concentrations (2, 20, 500 and 5000 μg/kg) were in a range from 76.7 to 104.9%. Results showed that the proposed method was simple, fast, economical and effective for the determination of RB in chili powder and chili oil. Considering the excellent sorptive performance of PCX for RB, further work should be done to evaluate the usefulness of the PCX in r-dSPE for the clean-up and analyses of other trace-level alkaline contaminants.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Rhodamine B, analytical standard
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamine B, suitable for fluorescence
Supelco
Methanol, analytical standard
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%