Merck
  • Metabolomic profiling of serum in the progression of Alzheimer's disease by capillary electrophoresis-mass spectrometry.
Metabolomic profiling of serum in the progression of Alzheimer's disease by capillary electrophoresis-mass spectrometry.
Electrophoresis (2014-08-20)
Raúl González-Domínguez, Antonia García, Tamara García-Barrera, Coral Barbas, José Luis Gómez-Ariza
ABSTRACT
There is high interest in the discovery of early diagnostic biomarkers of Alzheimer's disease, for which metabolomics exhibits a great potential. In this work, a metabolomic approach based on ultrafiltration and analysis by CE-MS has been used to obtain representative fingerprints of polar metabolites from serum samples in order to distinguish between patients with Alzheimer's disease, mild cognitive impairment, and healthy controls. By the use of partial least squares discriminant analysis it was possible to classify patients according to the disease stage and then identify potential markers. Significant increase was observed with progression of disease in levels of choline, creatinine, asymmetric dimethyl-arginine, homocysteine-cysteine disulfide, phenylalanyl-phenylalanine, and different medium chain acylcarnitines. On the other hand, asparagine, methionine, histidine, carnitine, acetyl-spermidine, and C5-carnitine were reduced in these serum samples. In this way, multiple essential pathways were found implicated in the underlying pathology, such as oxidative stress or defects in energy metabolism. However, the most interesting results are related to the association of several vascular risk factors with Alzheimer's disease.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Nα-Acetyl-L-ornithine
Sigma-Aldrich
Phe-Phe
Sigma-Aldrich
Purine, 98%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Choline chloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
DL-Cysteine, technical grade
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Methionine, European Pharmacopoeia (EP) Reference Standard
Histidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%