Merck
  • Ultrahigh-performance liquid chromatography-ion trap mass spectrometry characterization of the steroidal saponins of Dioscorea panthaica Prain et Burkill and its application for accelerating the isolation and structural elucidation of steroidal saponins.
Ultrahigh-performance liquid chromatography-ion trap mass spectrometry characterization of the steroidal saponins of Dioscorea panthaica Prain et Burkill and its application for accelerating the isolation and structural elucidation of steroidal saponins.
Steroids (2015-01-13)
Weihao Wang, Ye Zhao, Wenguang Jing, Jun Zhang, Hui Xiao, Qin Zha, An Liu
ABSTRACT
Dioscorea panthaica is a traditional Chinese medicinal herb used in the treatment of various physiological conditions, including cardiovascular disease, gastropathy and hypertension. Steroidal saponins (SS) are the main active ingredients of this herb and have effects on myocardial ischemia and cancer. The phytochemical evaluation of SS is both time-consuming and laborious, and the isolation and structural determination steps can be especially demanding. For this reason, the development of new methods to accelerate the processes involved in the identification, isolation and structural elucidation of SS is highly desirable. In this study, a new ultrahigh performance liquid chromatography-ion trap mass spectrometry (UHPLC-IT/MS(n)) method has been developed for the identification of the SS in D. panthaica Prain et Burkill. Notably, the current method can distinguish between spirostanol and furostanol-type compounds based on the fragmentation patterns observed by electrospray ionization-ion trap mass spectrometry (ESI-IT/MS(n)) analysis. UHPLC-IT/MS(n) was used to conduct a detailed investigation of the number, structural class and order of the sugar moieties in the sugar chains of the SS present in D. panthaica. The established fragmentation features were used to analyze the compounds found in the 65% ethanol fraction of the water extracts of D. panthaica. Twenty-three SS were identified, including 11 potential new compounds and six groups of isomers. Two of these newly identified SS were selected as representative examples, and their chemical structures were confirmed by (1)H and (13)C NMR analyses. This newly developed UHPLC-IT/MS(n) method therefore allowed for the efficient identification, isolation and structural determination of the SS in D. panthaica.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%