Merck
  • Organic anion transporting polypeptides and organic cation transporter 1 contribute to the cellular uptake of the flavonoid quercetin.
Organic anion transporting polypeptides and organic cation transporter 1 contribute to the cellular uptake of the flavonoid quercetin.
Naunyn-Schmiedeberg's archives of pharmacology (2014-06-21)
Hartmut Glaeser, Krystyna Bujok, Ingrid Schmidt, Martin F Fromm, Kathrin Mandery
ABSTRACT
Flavonoids such as quercetin and kaempferol mediate several health protective effects, e.g., anticancer effects. They are inhibitors of organic anion transporting polypeptides (OATP) and organic cation transporters (e.g., OCT2). However, little is known whether such transporters contribute to the cellular uptake of flavonoids. Therefore, we investigated the cellular uptake of kaempferol and quercetin using HEK293 cell lines stably expressing different human OATPs or OCT1. Kaempferol was not a substrate of any of the investigated transporters (OATP1A2, OATP1B1, OATP1B3, OATP2A1, OATP2B1, OATP3A1, OATP4A1, OATP5A1, and OCT1). Quercetin showed a significantly higher uptake into the HEK293-OATP1A2, HEK293-OATP2A1, HEK293-OATP2B1, and HEK293-OCT1 cells compared to control cells. The OATP1A2-, OATP2B1-, and OCT1-mediated quercetin uptake was inhibited by known inhibitors such as naringin, cyclosporin A, and quinidine, respectively. The cellular accumulation of quercetin into HEK293-OATP2A1 cells was not inhibited by prostaglandin E2 and diclofenac. The ionophore carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone (FCCP) reduced the net uptake of quercetin by increasing the uptake in the HEK293-control cells and causing no significant change in the HEK293-OATP2B1 cells indicating that quercetin follows the FCCP-driven proton flux through the plasma membrane. In addition to passive diffusion, the SLC transporters OATP1A2, OATP2B1, and OCT1 contribute to cellular accumulation of quercetin.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Quinidine, anhydrous
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium butyrate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Naringin, analytical standard
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Naringin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic