Merck
  • Prevention of hepatitis C virus infection by adoptive allogeneic immunotherapy using suicide gene-modified lymphocytes: an in vitro proof-of-concept.
Prevention of hepatitis C virus infection by adoptive allogeneic immunotherapy using suicide gene-modified lymphocytes: an in vitro proof-of-concept.
Gene therapy (2014-11-14)
C Leboeuf, J Roser-Schilder, M Lambotin, S Durand, T Wu, C Fauvelle, B Su, E Bôle-Richard, M Deschamps, C Ferrand, P Tiberghien, P Pessaux, T F Baumert, E Robinet
ABSTRACT
Hepatitis C virus (HCV)-induced, end-stage liver disease is a major indication for liver transplantation, but systematic graft reinfection accelerates liver disease recurrence. Transplantation recipients may be ineligible for direct-acting antivirals, owing to toxicity, resistance or advanced liver disease. Adoptive immunotherapy with liver graft-derived, ex vivo-activated lymphocytes was previously shown to prevent HCV-induced graft reinfections. Alternatively, the applicability and therapeutic efficacy of adoptive immunotherapy may be enhanced by 'ready for use' suicide gene-modified lymphocytes from healthy blood donors; moreover, conditional, prodrug-induced cell suicide may prevent potential side effects. Here, we demonstrate that allogeneic suicide gene-modified lymphocytes (SGMLs) could potently, dose- and time-dependently, inhibit viral replication. The effect occurs at effector:target cell ratios that exhibits no concomitant cytotoxicity toward virus-infected target cells. The effect, mediated mostly by CD56+ lymphocytes, is interleukin-2-dependent, IFN-γ-mediated and, importantly, resistant to calcineurin inhibitors. Thus, post-transplant immunosuppression may not interfere with this adoptive cell immunotherapy approach. Furthermore, these cells are indeed amenable to conditional cell suicide; in particular, the inducible caspase 9 suicide gene is superior to the herpes simplex virus thymidine kinase suicide gene. Our data provide in vitro proof-of-concept that allogeneic, third-party, SGMLs may prevent HCV-induced liver graft reinfection.
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard