Merck
  • Sequential immune monitoring in patients with melanoma and renal cell carcinoma treated with high-dose interleukin-2: immune patterns and correlation with outcome.
Sequential immune monitoring in patients with melanoma and renal cell carcinoma treated with high-dose interleukin-2: immune patterns and correlation with outcome.
Cancer immunology, immunotherapy : CII (2014-09-11)
David M Foureau, Asim Amin, Richard L White, William Anderson, Chase P Jones, Terry Sarantou, Iain H McKillop, Jonathan C Salo
ABSTRACT
Interleukin-2 (IL-2) therapy leads to clinically relevant responses in 10-16 % of patients with metastatic melanoma (MMEL) or 10-30 % of patients with metastatic renal cell carcinoma (MRCC). To date, no biomarkers have been validated to identify patients who are likely to respond. We hypothesized that changes in T cell subset distribution in patients undergoing IL-2 therapy may correlate with treatment outcomes. Immune profiles of 64 patients (27-MMEL, 37-MRCC) were evaluated using flow cytometry at baseline, during (≥three doses) and at the end of treatment cycle (30 ± 6 h after last dose), through two courses of IL-2 therapy. Changes in distribution and phenotype of circulating CD4 and CD8 lymphocyte subsets were compared (1) based on cancer types and (2) intra-patient during the course of the IL-2 therapy. Exploratory analysis of immunologic profiles was also performed based on treatment outcome. Independent of cancer type, IL-2 led to a transient decrease of circulating effector lymphocytes, while regulatory T cells gradually increased. Interleukin-2 differentially affected a subset of CD8 T cell expressing Foxp3, depending on malignancy type. In MMEL patients, IL-2 gradually expanded circulating CD8 Foxp3+ cells; in MRCC patients, IL-2 transiently increased expression of CD103 and CCR4 homing markers. Monitoring of adaptive immune variables early on and during the course of IL-2 therapy revealed transient alterations in immune profiles, specific to MMEL and MRCC patients, related to immune balance (and ultimately response to IL-2 therapy) or T cell egress from the circulation.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology