Merck
  • Pure and palladium-loaded Co3O4 hollow hierarchical nanostructures with giant and ultraselective chemiresistivity to xylene and toluene.
Pure and palladium-loaded Co3O4 hollow hierarchical nanostructures with giant and ultraselective chemiresistivity to xylene and toluene.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-02-26)
Su-Jin Hwang, Kwon-Il Choi, Ji-Wook Yoon, Yun Chan Kang, Jong-Heun Lee
ABSTRACT
Pure and palladium-loaded Co3O4 hollow hierarchical nanostructures consisting of nanosheets have been prepared by solvothermal self-assembly. The nanostructures exhibited an ultrahigh response and selectivity towards p-xylene and toluene. The responses (resistance ratio) of the palladium-loaded Co3O4 hollow hierarchical nanostructures to 5 ppm of p-xylene and toluene were as high as 361 and 305, respectively, whereas the selectivity values (response ratios) towards p-xylene and toluene over interference from ethanol were 18.1 and 16.1, respectively. We attributed the giant response and unprecedented high selectivity towards methylbenzenes to the abundant adsorption of oxygen by Co3O4, the high chemiresistive variation in the Co3O4 nanosheets (thickness≈11 nm), and the catalytic promotion of the specific gas-sensing reaction. The morphological design of the p-type Co3O4 nanostructures and loading of the palladium catalyst have paved a new way to monitoring the most representative indoor air pollutants in a highly selective, sensitive, and reliable manner.
Product Description
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
Cobalt(II) acetate, 99.99% trace metals basis
Supelco
Benzene, analytical standard
Supelco
Toluene, analytical standard
Supelco
p-Xylene, analytical standard
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Rhein, technical grade