Merck
  • Comparison of growth factor treatments on the fibrochondrogenic potential of canine fibroblast-like synoviocytes for meniscal tissue engineering.
Comparison of growth factor treatments on the fibrochondrogenic potential of canine fibroblast-like synoviocytes for meniscal tissue engineering.
Veterinary surgery : VS (2014-03-13)
Jason Spina, Jennifer Warnock, Katja Duesterdieck-Zellmer, Wendy Baltzer, Jesse Ott, Brian Bay
ABSTRACT
To determine the in vitro effects of differing growth factor treatments on the fibrochondrogenic potential of fibroblast-like synoviocytes from cruciate ligament deficient femorotibial joints of dogs. In vitro study. Synoviocytes from dogs (n = 8) with naturally occurring cruciate ligament insufficiency. Synoviocytes were cultured in monolayer and synthesized into tensioned synoviocyte bioscaffolds (TSB) suspended in media containing TGF-β3, or FGF-2, TGF-β1, and IGF-I. The 1,9-dimethylmethylene blue (DMMB) assay and toluidine blue stain assessed glycosaminoglycan content; hydroxyproline assay, and collagen I and II immunohistochemistry assessed collagen content. Biomechanical properties were determined by materials testing/force-deformation curves. All tissue cultures formed tensioned fibrous tissue-like constructs. Mean tissue cellularity and cellular viability was significantly greater in the triple growth factor-treated TSB by 0.09% and 44%, respectively. Percentage collagen content, and relative gene expression for collagen I, II, and aggrecan was not significantly different between groups. Median percentage of GAG content was significantly greater in triple growth factor-treated TSB by 1.6%. Biomechanical properties were not different in compression. Triple growth factor-treated TSB were significantly stronger in toughness, peak load to failure, and stiffness in tension. TGF-β3 cultured bioscaffolds failed to outperform triple growth factor-treated TSB. Architectural extracellular matrix (ECM) organization and cellularity likely explained the differences between groups. TGF-β3 alone cannot be recommended at this time for in vitro formation of autologous fibrocartilage bioscaffolds for meniscal deficiency.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material