Merck
  • Enteric-coating of pulsatile-release HPC capsules prepared by injection molding.
Enteric-coating of pulsatile-release HPC capsules prepared by injection molding.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-01-15)
E Macchi, L Zema, A Maroni, A Gazzaniga, L A Felton
ABSTRACT
Capsular devices based on hydroxypropyl cellulose (Klucel® LF) intended for pulsatile release were prepared by injection molding (IM). In the present work, the possibility of exploiting such capsules for the development of colonic delivery systems based on a time-dependent approach was evaluated. For this purpose, it was necessary to demonstrate the ability of molded cores to undergo a coating process and that coated systems yield the desired performance (gastric resistance). Although no information was available on the coating of IM substrates, some issues relevant to that of commercially-available capsules are known. Thus, preliminary studies were conducted on molded disks for screening purposes prior to the spray-coating of HPC capsular cores with Eudragit® L 30 D 55. The ability of the polymeric suspension to wet the substrate, spread, start penetrating and initiate hydration/swelling, as well as to provide a gastroresistant barrier was demonstrated. The coating of prototype HPC capsules was carried out successfully, leading to coated systems with good technological properties and able to withstand the acidic medium with no need for sealing at the cap/body joint. Such systems maintained the original pulsatile release performance after dissolution of the enteric film in pH 6.8 fluid. Therefore, they appeared potentially suitable for the development of a colon delivery platform based on a time-dependent approach.
Product Description
Supelco
Triethyl citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethyl citrate, ≥98.0% (GC)
Sigma-Aldrich
Triethyl citrate, ≥99%, FCC, FG
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Fluorescein sodium salt, p.a.
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
Fluorescein sodium salt, analytical standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material