Merck
  • Influence of dietary tender cluster beans (Cyamopsis tetragonoloba) on biliary proteins, bile acid synthesis and cholesterol crystal growth in rat bile.
Influence of dietary tender cluster beans (Cyamopsis tetragonoloba) on biliary proteins, bile acid synthesis and cholesterol crystal growth in rat bile.
Steroids (2014-12-24)
Chikkanna K Raghavendra, Krishnapura Srinivasan
ABSTRACT
Tender cluster beans (CBs; Cyamopsis tetragonoloba) are observed to possess anti-lithogenic potential in experimental mice. Formation of cholesterol gallstones in gallbladder is controlled by procrystallizing and anticrystallizing factors present in bile in addition to supersaturation of cholesterol. This study aimed at evaluating the influence of CB on biliary glycoproteins, low molecular weight (LMW) and high molecular weight (HMW) proteins, cholesterol nucleation time, and cholesterol crystal growth in rat hepatic bile. Groups of rats were fed for 10 weeks with 0.5% cholesterol to render the bile lithogenic. Experimental dietary interventions were: 10% freeze-dried CB, 1% garlic powder or their combination. Incorporation of CB into HCD decreased the cholesterol saturation index in bile, increased bile flow and biliary glycoproteins. Dietary CB prolonged cholesterol nucleation time in bile. Electrophoresis of biliary proteins showed the presence of high concentration of 27 kDa protein which might be responsible for the prolongation of cholesterol nucleation time in the CB fed group. Proteins of 20 kDa and 18 kDa were higher in CB treated animals, while the same were less expressed in HCD group. Biliary proteins from CB fed animals reduced cholesterol crystal growth index which was elevated in the presence of proteins from HCD group. Cholesterol-7α-hydroxylase and cholesterol-27-hydroxylase mRNA expression was increased in CB treated animals contributing to the bile acid synthesis. Thus, the beneficial anti-lithogenic effect of dietary CB which primarily is due to reduced cholesterol saturation index was additionally affected through a modulation of the nucleating and anti-nucleating proteins that affect cholesterol crystallization.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Triolein, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%