Merck
  • Glycaemic control is harder to achieve than blood pressure or lipid control in Irish adults with type 1 diabetes.
Glycaemic control is harder to achieve than blood pressure or lipid control in Irish adults with type 1 diabetes.
Diabetes research and clinical practice (2014-12-03)
T G Cotter, S F Dinneen, D A Healy, M J Bell, A Cunningham, P M O'Shea, F Dunne, T O'Brien, F M Finucane
ABSTRACT
We sought to determine the attainment of targets for glycaemic control and vascular risk reduction in an Irish cohort of T1DM adults. Of 797 patients (53% male, mean age 40.3 ± 14.8 years, HbA1c 8.5 ± 1.6% (69.6 ± 17.8 mmol mol(-1))), 15%, 68% and 62% achieved targets for HbA1c, blood pressure and LDL cholesterol, respectively.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications