Merck
  • Immunoprotection against influenza virus H9N2 by the oral administration of recombinant Lactobacillus plantarumNC8 expressing hemagglutinin in BALB/c mice.
Immunoprotection against influenza virus H9N2 by the oral administration of recombinant Lactobacillus plantarumNC8 expressing hemagglutinin in BALB/c mice.
Virology (2014-08-02)
Shao-Hua Shi, Wen-Tao Yang, Gui-Lian Yang, Yan-Long Cong, Hai-Bin Huang, Qian Wang, Ruo-Peng Cai, Li-Ping Ye, Jing-Tao Hu, Jing-Yu Zhou, Chun-Feng Wang, Yu Li
ABSTRACT
The H9N2 avian influenza virus (AIV) has become increasingly concerning due to its role in severe economic losses in the poultry industry. Transmission of AIV to mammals, including pigs and humans, has accelerated efforts to devise preventive strategies. To develop an effective oral vaccine against H9N2 AIV, a recombinant Lactobacillus plantarum NC8 strain expressing the hemagglutinin (HA) gene of H9N2 AIV was constructed in this study. Mice were orally immunized with the recombinant NC8-pSIP409-HA strain, and sIgA, IgG and HI antibodies were produced by the NC8-pSIP409-HA strain, which also induced CD8(+) T cell immune responses. Most importantly, oral administration produced complete protection against challenge with mouse-adapted H9N2 virus. These results indicate that the recombinant NC8-pSIP409-HA was more effective at inducing the mucosal, humoral and cellular immune responses. Therefore, L. plantarum NC8-pSIP409-HA could become a promising oral vaccine candidate against H9N2 AIV.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)