Merck
  • Crotoxin suppresses the tumorigenic properties and enhances the antitumor activity of Iressa® (gefinitib) in human lung adenocarcinoma SPCA‑1 cells.
Crotoxin suppresses the tumorigenic properties and enhances the antitumor activity of Iressa® (gefinitib) in human lung adenocarcinoma SPCA‑1 cells.
Molecular medicine reports (2014-10-14)
Junhua Wang, Xiuguang Qin, Zhiqiang Zhang, Meiling Chen, Ying Wang, Baoqin Gao
ABSTRACT
Crotoxin (CrTX) is a neurotoxin isolated from the venom of the South American rattlesnake. Previous studies demonstrated that CrTX was able to inhibit the activity of the growth factor receptor tyrosine kinase and that CrTX possesses potent antitumor activity when combined with Iressa, an epidermal growth factor receptor inhibitor. The aim of the present study was to determine the antitumor effect of CrTX and the combination of CrTX with Iressa in lung adenocarcinoma SPCA‑1 cells. The results demonstrated that CrTX inhibited the cellular growth of SPCA‑1 cells via G1 arrest and induction of apoptosis. In addition, the c‑Jun N‑terminal kinase pathway was important in CrTX‑induced apoptosis in SPCA‑1 cells. Notably, CrTX was able to significantly enhance the cytotoxic effects of Iressa in SPCA‑1 cells. The in vivo antitumor assay demonstrated that treatment with either CrTX or Iressa significantly inhibited tumor growth, while the combination of CrTX and Iressa demonstrated the most significant antitumor activity, which was reflected by tumor weight and angiogenesis, presented as microvascular density. Therefore, the present study suggested that CrTX is a potential anti‑lung cancer agent and sensitizer of Iressa.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC