Merck
  • Association of reduced folate carrier-1 (RFC-1) polymorphisms with ischemic stroke and silent brain infarction.
Association of reduced folate carrier-1 (RFC-1) polymorphisms with ischemic stroke and silent brain infarction.
PloS one (2015-02-07)
Yunkyung Cho, Jung O Kim, Jeong Han Lee, Hye Mi Park, Young Joo Jeon, Seung Hun Oh, Jinkun Bae, Young Seok Park, Ok Joon Kim, Nam Keun Kim
ABSTRACT
Stroke is the second leading cause of death in the world and in South Korea. Ischemic stroke and silent brain infarction (SBI) are complex, multifactorial diseases influenced by multiple genetic and environmental factors. Moderately elevated plasma homocysteine levels are a major risk factor for vascular diseases, including stroke and SBI. Folate and vitamin B12 are important regulators of homocysteine metabolism. Reduced folate carrier (RFC), a bidirectional anion exchanger, mediates folate delivery to a variety of cells. We selected three known RFC-1 polymorphisms (-43C>T, 80A>G, 696T>C) and investigated their relationship to cerebral infarction in the Korean population. We used the polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP) method to analyze associations between the three RFC-1 polymorphisms, disease status, and folate and homocysteine levels in 584 ischemic stroke patients, 353 SBI patients, and 505 control subjects. The frequencies of the RFC-1 -43TT, 80GG, and 696CC genotypes differed significantly between the stroke and control groups. The RFC-1 80A>G substitution was also associated with small artery occlusion and SBI. In a gene-environment analysis, the RFC-1 -43C>T, 80A>G, and 696T>C polymorphisms in the ischemic stroke group had combined effects with all environmental factors. In summary, we found that the RFC-1 -43C>T, 80A>G, and 696T>C polymorphisms may be risk factors for ischemic stroke.
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Supelco
DL-Homocysteine, analytical standard