Merck
  • Pharmaceutical properties of a low-substituted hydroxypropyl cellulose (L-HPC) hydrogel as a novel external dressing.
Pharmaceutical properties of a low-substituted hydroxypropyl cellulose (L-HPC) hydrogel as a novel external dressing.
International journal of pharmaceutics (2014-12-03)
Atsushi Ogawa, Sachie Nakayama, Mami Uehara, Yasuhiro Mori, Mai Takahashi, Tetsuya Aiba, Yuji Kurosaki
ABSTRACT
Controlling the moisture balance between exudates and their transpiration from the surface of wounded skin is important for healing. Low-substituted hydroxypropyl cellulose (L-HPC) hydrogel sheets (HGSs) possessing high water retention and water vapor transmission properties were prepared by neutralizing the highly viscous alkaline liquid of 7-10% L-HPC. Glycerol-impregnated L-HPC hydrogel sheets (L-HPC G-HGSs) were obtained by exchanging aqueous liquid in L-HPC HGSs. The physical characteristics required for wound dressings, i.e., mechanical strength, adhesive strength, and water retention properties, as well as the water vapor transmission (WVT) properties of L-HPC HGSs and L-HPC G-HGSs were evaluated. The mechanical strengths of L-HPC HGSs were enhanced with increases in the L-HPC content. The impregnation of glycerol in L-HPC HGSs yielded a significantly elasticated sheet. The adhesive strengths of L-HPC HGSs were significantly lower than those of commercial medical dressings. Water retention in L-HPC HGSs after being stored for 2h at 37°C was approximately 50%. The WVT rate of 7% L-HPC HGS was approximately 40g/m(2)/h, which was markedly higher than that of silicone gel type medical dressings. In conclusion, L-HPC HGSs are promising dressings that maintain an adequate moisture balance by transpiring excessive wound exudates with less damage to the healing wound.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M