Merck
  • Si/SiO2 quantum dots cause cytotoxicity in lung cells through redox homeostasis imbalance.
Si/SiO2 quantum dots cause cytotoxicity in lung cells through redox homeostasis imbalance.
Chemico-biological interactions (2014-07-06)
Miruna S Stan, Indira Memet, Cornelia Sima, Traian Popescu, Valentin S Teodorescu, Anca Hermenean, Anca Dinischiotu
ABSTRACT
Si/SiO2 quantum dots (QDs) are novel particles with unique physicochemical properties that promote them as potential candidates for biomedical applications. Although their interaction with human cells has been poorly investigated, oxidative stress appears to be the main factor involved in the cytotoxicity of these nanoparticles. In this study, we show for the first time the influence of Si/SiO2 QDs on cellular redox homeostasis and glutathione distribution in human lung fibroblasts. The nanoparticles morphology, composition and structure have been investigated using high resolution transmission electron microscopy (HRTEM), selected area electron diffraction (SAED), energy-dispersive X-ray spectroscopy (EDX) and X-ray diffraction (XRD) analysis. MRC-5 cells (human lung fibroblasts) were incubated with various concentrations of Si/SiO2 QDs ranging between 25 and 200 μg/mL for up to 72 h. The results of the MTT and sulforhodamine B assays showed that exposure to QDs led to a time-dependent decrease in cell viability and biomass. The increase in reactive oxygen species (ROS) and malondialdehyde (MDA) levels together with the lower glutathione content suggested that the cellular redox homeostasis was altered. Regarding GSH distribution, the first two days of treatment resulted in a localization of GSH mainly in the cytoplasm, while at longer incubation time the nuclear/cytoplasmic ratio indicated a nuclear localization. These modifications of cell redox state also affected the redox status of proteins, which was demonstrated by the accumulation of oxidized proteins and actin S-glutathionylation. In addition, the externalization of phosphatidylserine provided evidence that apoptosis might be responsible for cell death, but necrosis was also revealed. Our results suggest that Si/SiO2 quantum dots exerted cytotoxicity on MRC-5 cells by disturbing cellular homeostasis which had an effect upon protein redox status.
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized, ≥98%, lyophilized powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Glutathione oxidized
SAFC
Sodium deoxycholate
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %