Merck
  • Targeted antioxidant treatment decreases cardiac alternans associated with chronic myocardial infarction.
Targeted antioxidant treatment decreases cardiac alternans associated with chronic myocardial infarction.
Circulation. Arrhythmia and electrophysiology (2014-12-11)
Bradley N Plummer, Haiyan Liu, Xiaoping Wan, Isabelle Deschênes, Kenneth R Laurita
ABSTRACT
In myocardial infarction (MI), repolarization alternans is a potent arrhythmia substrate that has been linked to Ca2+ cycling proteins, such as sarcoplasmic reticulum Ca2+ ATPase (SERCA2a), located in the sarcoplasmic reticulum. MI is also associated with oxidative stress and increased xanthine oxidase (XO) activity, an important source of reactive oxygen species (ROS) in the sarcoplasmic reticulum that may reduce SERCA2a function. We hypothesize that in chronic MI, XO-mediated oxidation of SERCA2a is a mechanism of cardiac alternans. Male Lewis rats underwent ligation of the left anterior descending coronary artery (n=54) or sham procedure (n=24). At 4 weeks, optical mapping of intracellular Ca2+ and ROS was performed. ECG T-wave alternans (ECG ALT) and Ca2+ transient alternans (Ca2+ALT) were induced by rapid pacing (300-120 ms) before and after the XO inhibitor allopurinol (ALLO, 50 µmol/L). In MI, ECG ALT (2.32±0.41%) and Ca2+ ALT (22.3±4.5%) were significantly greater compared with sham (0.18±0.08%, P<0.001; 0.79±0.32%, P<0.01). Additionally, ROS was increased by 137% (P<0.01) and oxidation of SERCA2a by 30% (P<0.05) in MI compared with sham. Treatment with ALLO significantly decreased ECG ALT (-77±9%, P<0.05) and Ca2+ ALT (-56±7%, P<0.05) and, importantly, reduced ROS (-65%, P<0.01) and oxidation of SERCA2a (-38%, P<0.05). CaMKII inhibition and general antioxidant treatment had no effect on ECG ALT and Ca2+ ALT. These results demonstrate, for the first time, that in MI, increased ROS from XO is a significant cause of repolarization alternans. This suggests that targeting XO ROS production may be effective at preventing arrhythmia substrates in chronic MI.
Sigma-Aldrich
Bromobimane, suitable for fluorescence, BioReagent, ≥95% (HPCE)
Sigma-Aldrich
Bromobimane, ≥97% (HPLC)
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Allopurinol, European Pharmacopoeia (EP) Reference Standard
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material