Merck
  • Qualitative and spatial metabolite profiling of lichens by a LC-MS approach combined with optimised extraction.
Qualitative and spatial metabolite profiling of lichens by a LC-MS approach combined with optimised extraction.
Phytochemical analysis : PCA (2014-08-19)
Delphine Parrot, Tiphaine Peresse, Eric Hitti, Daniel Carrie, Martin Grube, Sophie Tomasi
ABSTRACT
Lichens are self-sustaining partnerships comprising fungi as shape-forming partners for their enclosed symbiotic algae. They produce a tremendous diversity of metabolites (1050 metabolites described so far). A comparison of metabolic profiles in nine lichen species belonging to three genera (Lichina, Collema and Roccella) by using an optimised extraction protocol, determination of the fragmentation pathway and the in situ localisation for major compounds in Roccella species. Chemical analysis was performed using a complementary study combining a Taguchi experimental design with qualitative analysis by high-performance liquid chromatography coupled with mass spectrometry techniques. Optimal conditions to obtain the best total extraction yield were determined as follows: mortar grinding to a fine powder, two successive extractions, solid:liquid ratio (2:60) and 700 rpm stirring. Qualitative analysis of the metabolite profiling of these nine species extracted with the optimised method was corroborated using MS and MS/MS approaches. Nine main compounds were identified: 1 β-orcinol, 2 orsellinic acid, 3 putative choline sulphate, 4 roccellic acid, 5 montagnetol, 6 lecanoric acid, 7 erythrin, 8 lepraric acid and 9 acetylportentol, and several other compounds were reported. Identification was performed using the m/z ratio, fragmentation pathway and/or after isolation by NMR analysis. The variation of the metabolite profile in differently organised parts of two Roccella species suggests a specific role of major compounds in developmental stages of this symbiotic association. Metabolic profiles represent specific chemical species and depend on the extraction conditions, the kind of the photobiont partner and the in situ localisation of major compounds.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Cyclohexane, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material