Merck
  • Determination of acacetin in rat plasma by UPLC-MS/MS and its application to a pharmacokinetic study.
Determination of acacetin in rat plasma by UPLC-MS/MS and its application to a pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-24)
Li-hua Fan, Xiaoheng Li, De-yuan Chen, Ning Zhang, Yiyan Wang, Yuanyuan Shan, Yuanyuan Hu, Ren-ai Xu, Jian Jin, Ren-Shan Ge
ABSTRACT
A rapid, sensitive and selective ultra-performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS) was developed and validated for the determination and pharmacokinetic investigation of acacetin in rat plasma. Sample preparation was accomplished through a simple one-step deproteinization procedure with 0.2 mL of acetonitrile to a 0.1 mL plasma sample. Plasma samples were separated by UPLC on an Acquity UPLC BEH C18 column using a mobile phase consisting of acetonitrile-0.1% formic acid in water with gradient elution. The total run time was 2.0 min and the elution of acacetin was at 0.83 min. The detection was performed on a triple quadrupole tandem mass spectrometer equipped with positive-ion electrospray ionization (ESI) by multiple reaction monitoring (MRM) of the transitions at m/z 285.3→242.2 for acacetin and m/z 237.2→194.3 for carbamazepine (internal standard). The calibration curve was linear over the range of 1-1600 ng/mL with a lower limit of quantitation (LLOQ) of 1.0 ng/mL. Mean recovery of acacetin in plasma was in the range of 78.4-85.2%. Intra-day and inter-day precision were both <10.5%. This method was successfully applied in pharmacokinetic study after intravenous administration of 5.0mg/kg acacetin in rats.
Product Description
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Carbamazepine, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard