Merck
Microchip-based human serum atherogenic lipoprotein profile analysis.
Analytical biochemistry (2014-09-15)
Hua Wang, Wei Zhang, Jun Wan, Weiwei Liu, Bo Yu, Qinghui Jin, Ming Guan
ABSTRACT
Owing to the mounting evidence of serum lipid changes in atherosclerosis, there has been increasing interest in developing new methods for analyzing atherogenic lipoprotein profiles. The separation of lipoprotein and lipoprotein subclasses has been demonstrated using a microchip capillary electrophoresis (CE) system [Chromatographia 74 (2011) 799-805]. In contrast to this previous study, the current report demonstrates that sdLDL peak efficiencies can be improved dramatically by adding gold nanoparticles (AuNPs) to the sample. Moreover, NBD C6-ceramide was identified as a satisfactory dye for specific labeling and quantitation of individual serum lipoproteins. The accuracy of the method was evaluated by comparison with ultracentrifuge separated small, dense, low-density lipoprotein (sdLDL). A high correlation was observed between these two methods for sdLDL cholesterol. Lipid levels were investigated between atherosclerotic patients and healthy controls. The variation of serum atherogenic lipoprotein profiles for atherosclerotic patients pre- and post-treatment was assessed by microchip CE. This method has potential for the rapid and sensitive detection of different lipoprotein classes as well as their subclasses and, therefore, is suitable for routine clinical applications. Microchip-based atherogenic lipoprotein profile assays will greatly improve the analysis of risk factors in atherosclerosis and will provide useful information for monitoring the effect of therapies on atherosclerotic disease.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
SAFC
Tricine
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethylene glycol 5 M solution