Merck
  • Encapsulation of porcine pancreatic islets within an immunoprotective capsule comprising methacrylated glycol chitosan and alginate.
Encapsulation of porcine pancreatic islets within an immunoprotective capsule comprising methacrylated glycol chitosan and alginate.
Journal of biomedical materials research. Part B, Applied biomaterials (2014-06-12)
Anna Louise Hillberg, Matthew Oudshoorn, Janice B B Lam, Kalyani Kathirgamanathan
ABSTRACT
Encapsulation of cells in biocompatible polymer matrices represents a powerful tool for cell-based therapies and therapeutic delivery systems. This technology has successfully been used to deliver pancreatic islets to humans for the treatment of Type 1 diabetes. However, the clinical impact of this technology may be improved by reducing the inflammatory response brought on after implantation of capsules in vivo. Within this study a biocompatible polymeric delivery system combining alginate and photo-crosslinked methacrylated glycol chitosan (MGC) was developed. This approach involved encapsulating cells in calcium-alginate beads, coating with MGC and photo-polymerizing using UVA in the presence of photo-initiator (VA-086), resulting in the formation of capsules ∼600 µm in size. Crosslinking of the MGC outer wall allowed control over capsule swelling and improved the capsules overall properties. Capsule characterization demonstrated the stabilizing influence of polymerization and fluorescence imaging showed that the distribution of glycol chitosan is dependent on molecular weight. Good islet viability and insulin release was demonstrated in vitro over the course of a month, and in vivo transplantation of the capsules demonstrated good biocompatibility, particularly when compared with standard alginate/poly-l-ornithine/alginate capsules.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Theophylline, United States Pharmacopeia (USP) Reference Standard
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder