Merck
  • Safety evaluation of standardized allergen extract of Japanese cedar pollen for sublingual immunotherapy.
Safety evaluation of standardized allergen extract of Japanese cedar pollen for sublingual immunotherapy.
Regulatory toxicology and pharmacology : RTP (2015-02-24)
Yuko Mitobe, Yasuki Yokomoto, Katsuyo Ohashi-Doi
ABSTRACT
Japanese cedar (JC) pollinosis is caused by Japanese cedar pollen (JCP) and most common seasonal allergic disease in Japan. Subcutaneous immunotherapy (SCIT) with allergen extract of JCP (JCP-allergen extract) is well established for JC pollinosis treatment with improvement of symptoms. However, major drawbacks for SCIT are repeated painful injections, frequent hospital visits and anaphylactic risk. Currently, sublingual immunotherapy (SLIT) has received much attention as an advanced alternative application with lower incidence of systemic reactions because the liquid or tablet form of allergen is placed under the tongue. The aim of this study was safety evaluation of standardized JCP-allergen extract currently developed for SLIT in JC pollinosis. JCP-allergen extract showed no potential genotoxicity. No systemic effects were observed in rats administered JCP-allergen extract orally for 26 weeks followed by 4-week recovery period. Mild local reactions such as hyperplasia and increased globule leukocytes resulting from vehicle (glycerin)-induced irritation were observed in stomach. No-observed-adverse-effect level was greater than 10,000 JAU/kg/day for systemic toxicity, equivalent to 300-fold the human dose. No local irritation was found in rabbits oral mucosae by 7-day sublingual administration. These results demonstrate the safe profile of standardized JCP-allergen extract, suggesting it is suitable for SLIT in JC pollinosis.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard