Merck
  • The transcriptional signatures of cells from the human Peyronie's disease plaque and the ability of these cells to generate a plaque in a rat model suggest potential therapeutic targets.
The transcriptional signatures of cells from the human Peyronie's disease plaque and the ability of these cells to generate a plaque in a rat model suggest potential therapeutic targets.
The journal of sexual medicine (2014-12-17)
Robert A Gelfand, Dolores Vernet, Istvan Kovanecz, Jacob Rajfer, Nestor F Gonzalez-Cadavid
ABSTRACT
The success of medical therapies for Peyronie's disease (PD) has not been optimal, possibly because many of them went directly to clinical application without sufficient preclinical scientific research. Previous studies revealed cellular and molecular pathways involved in the formation of the PD plaque and in particular the role of the myofibroblast. The current work aimed to determine under normal and fibrotic conditions what differentiates PD cells from tunica albuginea (TA) and corpora cavernosa (CC) cells by defining their global transcriptional signatures and testing in vivo whether PD cells can generate a PD-like plaque. Human TA, PD, and CC cells were grown with transforming growth factor beta 1 (TGFβ1; TA+, PD+, CC+) or without it (TA-, PD-, CC-) and assayed by (i) immunofluorescence, Western blot and RT-PCR for myofibroblast, smooth muscle cell and stem cell markers; (ii) collagen content; and (iii) DNA microarray analysis. The ability of PD+ cells to induce a PD-like plaque in an immuno-suppressed rat model was assessed by Masson trichrome and Picrosirius Red stainings. Fibroproliferative features of PD cells and identification of related key genes as novel targets to reduce plaque size. Upon TGFβ1stimulation, collagen levels were increased by myofibroblasts in the PD+ but not in the CC+ cells. The transcriptional signature of the PD- cells identified fibroproliferative, myogenic (myofibroblasts), inflammatory, and collagen turnover genes that differentiate them from TA- or CC- cells and respond to TGFβ1 with a PD+ fibrotic phenotype, by upregulation of IGF-1, ACTG2, MYF5, ACTC1, PSTN, COL III, MMP3, and others. The PD+ cells injected into the TA of the rat induce a PD-like plaque. This suggests a novel combination therapy to eliminate a PD plaque by targeting the identified genes to (i) improve collagenase action by stimulating endogenous metalloproteinases specific to key collagen types and (ii) counteract fibromatosis by inhibiting myofibroblast generation, proliferation, and/or apoptosis.
Product Description
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SAFC
Glycine
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)