Merck
  • Vitamin D rescues dysfunction of fetal endothelial colony forming cells from individuals with gestational diabetes.
Vitamin D rescues dysfunction of fetal endothelial colony forming cells from individuals with gestational diabetes.
Placenta (2015-02-17)
J Gui, A Rohrbach, K Borns, P Hillemanns, L Feng, C A Hubel, F von Versen-Höynck
ABSTRACT
Gestational diabetes (GDM) is associated with long-term cardiovascular and metabolic diseases in offspring. However, the mechanisms are not well understood. We explored whether fetal exposure to a diabetic environment is associated with fetal endothelial progenitor cell dysfunction, and whether vitamin D can reverse the impairment. Nineteen women with uncomplicated pregnancies and 18 women with GDM were recruited before delivery. Time to first appearance of endothelial colony forming cell (ECFC) colonies and number of ECFC colonies formed from culture of cord peripheral blood mononuclear cells were determined. Angiogenesis-related functions of ECFCs in vitro were tested in the presence or absence of vitamin D. Fetal ECFCs from GDM pregnancies formed fewer colonies in culture (P = 0.04) and displayed reduced proliferation (P = 0.02), migration (P = 0.04) and tubule formation (P = 0.03) compared to uncomplicated pregnancies. Fetal ECFCs exposed to hyperglycemia in vitro exhibited less migration (P < 0.05) and less tubule formation (P < 0.05) than normoglycemic control. Vitamin D significantly improved the dysfunction of fetal ECFCs from pregnancies complicated by GDM or after exposure of healthy ECFCs to hyperglycemia. Fetal ECFCs from GDM pregnancies or ECFCs exposed to hyperglycemia in vitro exhibit reduced quantity and impaired angiogenesis-related functions. Vitamin D significantly rescues these functions. These findings may have implications for vascular function of infants exposed to a diabetic intrauterine environment.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Cholecalciferol, analytical standard
Supelco
D-(+)-Glucose, analytical standard
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications