Merck
  • GC-MS/MS and LC-MS/MS studies on unlabelled and deuterium-labelled oleic acid (C18:1) reactions with peroxynitrite (O=N-O-O⁻) in buffer and hemolysate support the pM/nM-range of nitro-oleic acids in human plasma.
GC-MS/MS and LC-MS/MS studies on unlabelled and deuterium-labelled oleic acid (C18:1) reactions with peroxynitrite (O=N-O-O⁻) in buffer and hemolysate support the pM/nM-range of nitro-oleic acids in human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-02-11)
Arne Trettin, Anke Böhmer, Alexander A Zoerner, Frank-Mathias Gutzki, Jens Jordan, Dimitrios Tsikas
ABSTRACT
Oleic acid (cis-9,10-octadecenoic acid) is the most abundant monounsaturated fatty acid in human blood. Peroxynitrite (ONOO(-)) is a short-lived species formed from the reaction of nitric oxide (NO) and superoxide (O2(-)). Peroxynitrite is a potent oxidizing and moderate nitrating agent. We investigated reactions of unlabelled and deuterium labelled oleic acid in phosphate buffered saline (PBS) and lysed human erythrocytes with commercially available sodium peroxynitrite (Na(+)ONOO(-)). Non-derivatized reaction products were analyzed by spectrophotometry, HPLC with UV absorbance detection, and LC-MS/MS electrospray ionization in the negative-ion mode. Reaction products were also analyzed by GC-MS/MS in the electron capture negative-ion chemical ionization mode after derivatization first with pentafluorobenzyl (PFB) bromide and then with N,O-bis(trimethylsilyl)trifluoroacetamide. Identified oleic acid reaction products in PBS and hemolysate include cis-9,10-epoxystearic acid and trans-9,10-epoxystearic acid (about 0.1% with respect to oleic acid), threo- and erythro-9,10-dihydroxy-stearic acids. Vinyl nitro-oleic acids, 9-nitro-oleic acid (9-NO2OA) and 10-nitro-oleic acid (10-NO2OA), or other nitro-oleic acids were not found to be formed from the reaction of oleic acid with peroxynitrite in PBS or hemolysate. Our in vitro study suggests that peroxynitrite oxidizes but does not nitrate oleic acid in biological samples. Unlike thiols and tyrosine, oleic acid is not susceptible to peroxynitrite. GC-MS/MS analysis of PFB esters is by far more efficient than LC-MS/MS analysis of non-derivatized oleic acid and its derivates. Our in vitro results support our previous in vivo findings that nitro-oleic acid plasma concentrations of healthy and diseased subjects are in the pM/nM-range.
Product Description
Supelco
Ethyl acetate, analytical standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzyl bromide, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Supelco
Acetonitrile, analytical standard
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%