Merck
Detection and in vitro metabolism of AOD9604.
Drug testing and analysis (2014-09-12)
Holly D Cox, Stacy J Smeal, Cole M Hughes, James E Cox, Daniel Eichner
ABSTRACT
AOD9604 is a peptide consisting of the C-terminal fragment of human growth hormone from amino acids 177-191 with an additional tyrosine residue at the N-terminus of the peptide. It is reported to mimic the lipolytic properties of growth hormone without the diabetogenic side effects. Therefore, AOD9604 may be used as a performance enhancing drug and is banned by the World Anti-doping Agency (WADA). The peptide is available on several Internet websites and was recently identified in confiscated vials in the USA. To detect abuse of the peptide in athletes, a solid-phase extraction method was validated in urine with a limit of detection of 50 pg/mL. The method has good linearity, precision (<20%), specificity and recovery (62%). Six potential metabolites of the peptide were identified after incubation of AOD9604 in serum and urine. Quantification of the metabolites in serum identified a single metabolite, consisting of amino acids CRSVEGSCG, which is significantly more stable than the other metabolites or the parent compound. Screening for AOD9604 and the stable metabolite may potentially allow an increased window of detection.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material