Merck
  • Dimethyl sulphoxide and Ca2+ stimulate assembly of Vibrio cholerae FtsZ.
Dimethyl sulphoxide and Ca2+ stimulate assembly of Vibrio cholerae FtsZ.
Biochimie (2014-06-28)
Abhisek Chatterjee, Gopal Chakrabarti
ABSTRACT
We cloned, overexpressed and purified Vibrio cholerae FtsZ protein for the first time. We used several complementary techniques to probe and compare the comparative assembly properties of recombinant Vibrio cholerae FtsZ (VcFtsZ) and Escherichia coli FtsZ (EcFtsZ). We observed that VcFtsZ polymerized at a slower rate than EcFtsZ and interestingly its polymerization was highly dependent on the presence of Ca(2+) ion. Furthermore, DMSO specifically modulated the polymerization of VcFtsZ, promoted polymer bundling and increased the stability of the VcFtsZ protofilaments. Whereas DMSO showed no significant stimulatory effect on the assembly and bundling of EcFtsZ. Transmission electron microscopy experiments demonstrated that in presence of 8% DMSO the average thickness of the VcFtsZ polymers were increased significantly. DMSO specifically stabilized the VcFtsZ polymers against dilution induced disassembly and it reduced the GTPase activity of VcFtsZ. These results collectively suggested that despite lot of sequence homology, the assembly of VcFtsZ and EcFtsZ are differently regulated processes. We expect to use this knowledge of assembly properties of VcFtsZ for screening of small molecules against VcFtsZ for development of anti-cholera agent.
Product Description
Supelco
trans-Sinapic acid, analytical standard
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Benzamidine, ≥95.0%
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%