Merck
  • Long circulating micelles of an amphiphilic random copolymer bearing cell outer membrane phosphorylcholine zwitterions.
Long circulating micelles of an amphiphilic random copolymer bearing cell outer membrane phosphorylcholine zwitterions.
Acta biomaterialia (2015-02-01)
Jing Zhao, Yu-Dong Chai, Jing Zhang, Peng-Fei Huang, Kenichi Nakashima, Yong-Kuan Gong
ABSTRACT
Polymeric micelles with cell outer membrane mimetic structure were prepared in water from amphiphilic random copolymers bearing both the hydrophilic phosphorylcholine zwitterions and hydrophobic octadecyl side chains of cell outer membrane. The polymeric micelles showed sizes ranging from 80 nm to 120 nm in hydrodynamic diameter and zeta-potentials from -6.4 mV to -2.4 mV by dynamic light scattering measurements. The micelles loaded with 6-coumarin as a fluorescence probe were stable to investigate their blood circulation and biodistribution. The in vitro phagocytosis results using murine peritoneal macrophages showed 10-fold reduction compared with a reference micelle. The in vivo blood circulation half-life of the polymeric micelles following intravenous administration in New Zealand Rabbits was increased from 0.55 h to 90.5h. More interestingly, tissue distribution results showed that the concentration of the micelles in the kidney is 4-fold higher than that in the liver and other organs 48 h after administration. The results of this work show great promise for designing more effective stealth drug carriers that can minimize reticuloendothelial system clearance and circulate for long time to reach target by using simple cell membrane mimetic random copolymer micelles.
Sigma-Aldrich
[2-(Methacryloyloxy)ethyl]trimethylammonium chloride solution, 75 wt. % in H2O
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Supelco
Methanol, analytical standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%