Merck
  • Combinatorial regulation of a signal-dependent activator by phosphorylation and acetylation.
Combinatorial regulation of a signal-dependent activator by phosphorylation and acetylation.
Proceedings of the National Academy of Sciences of the United States of America (2014-11-19)
Jose C Paz, Sangho Park, Naomi Phillips, Shigenobu Matsumura, Wen-Wei Tsai, Lawryn Kasper, Paul K Brindle, Guangtao Zhang, Ming-Ming Zhou, Peter E Wright, Marc Montminy
ABSTRACT
In the fasted state, increases in catecholamine signaling promote adipocyte function via the protein kinase A-mediated phosphorylation of cyclic AMP response element binding protein (CREB). CREB activity is further up-regulated in obesity, despite reductions in catecholamine signaling, where it contributes to the development of insulin resistance. Here we show that obesity promotes the CREB binding protein (CBP)-mediated acetylation of CREB at Lys136 in adipose. Under lean conditions, CREB acetylation was low due to an association with the energy-sensing NAD(+)-dependent deacetylase SirT1; amounts of acetylated CREB were increased in obesity, when SirT1 undergoes proteolytic degradation. Whereas CREB phosphorylation stimulated an association with the KIX domain of CBP, Lys136 acetylation triggered an interaction with the CBP bromodomain (BRD) that augmented recruitment of this coactivator to the promoter. Indeed, coincident Ser133 phosphorylation and Lys136 acetylation of CREB stimulated the formation of a ternary complex with the KIX and BRD domains of CBP by NMR analysis. As disruption of the CREB:BRD complex with a CBP-specific BRD inhibitor blocked effects of CREB acetylation on target gene expression, our results demonstrate how changes in nutrient status modulate cellular gene expression in response to hormonal signals.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium Fluoride Solution
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Pricing and availability is not currently available.
Sigma-Aldrich
Water, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Deionized
Pricing and availability is not currently available.
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pricing and availability is not currently available.
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pricing and availability is not currently available.
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pricing and availability is not currently available.