Merck
  • Protein S and factor V in regulation of coagulation on platelet microparticles by activated protein C.
Protein S and factor V in regulation of coagulation on platelet microparticles by activated protein C.
Thrombosis research (2014-05-20)
Sofia Somajo, Ruzica Livaja Koshiar, Eva Norström, Björn Dahlbäck
ABSTRACT
Platelets are the main source of microparticles in plasma and the concentration of microparticles is increased in many diseases. As microparticles expose negatively charged phospholipids, they can bind and assemble the procoagulant enzyme-cofactor complexes. Our aim was to elucidate possible regulation of these complexes on microparticles by the anticoagulant protein C system. Platelets were activated with thrombin ± collagen or the calcium ionophore A23187 ± thrombin to generate microparticles. The microparticles were analyzed using flow cytometry and functional coagulation assays to characterize parameters with importance for the activated protein C system. Activation with A23187+thrombin was most efficient, fully converting the platelets to microparticle-like vesicles, characterized by high lactadherin and protein S binding capacity. Suppression of thrombin generation by activated protein C in plasma spiked with these microparticles was dependent on the presence of plasma protein S. Experiments with purified components showed that activated protein C inhibited both factor Va and factor VIIIa on the microparticle surface. Inhibition of factor Va was stimulated by, but not fully dependent on, the presence of protein S. In the factor VIIIa-degradation, activated protein C was dependent on the addition of protein S, and exogenous factor V further increased the efficiency. Protein S is crucial for activated protein C-mediated inhibition of thrombin generation on platelet-derived microparticles in plasma. Moreover, protein S and factor V are synergistic cofactors in the inhibition of factor VIIIa. The results demonstrate that the activated protein C system has the capacity to counterbalance the procoagulant ability of microparticles.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)