Merck
  • P66Shc mediates increased platelet activation and aggregation in hypercholesterolemia.
P66Shc mediates increased platelet activation and aggregation in hypercholesterolemia.
Biochemical and biophysical research communications (2014-05-23)
Santosh Kumar, Ajit Vikram, Young-Rae Kim, Julia S Jacobs, Kaikobad Irani
ABSTRACT
Hypercholesterolemia leads to a prothrombotic phenotype. Platelet hyperactivity associated with hypercholesterolemia has been attributed, in part, to oxidative stress. P66Shc is a well-known determinant of cellular and organismal oxidative stress. However, its role in platelet biology is not known. We hypothesized that p66Shc mediates platelet hyperactivation and hyperaggregation in hypercholesterolemia. P66Shc was expressed in both human and mouse platelets, as determined by qRT-PCR and immunoblotting. Mouse platelet p66Shc expression was upregulated by hypercholesterolemia induced by high-fat diet feeding. Compared to wild-type mice, high-fat diet-induced p66Shc expression in platelets was suppressed in transgenic mice expressing a short hairpin RNA targeting p66Shc (p66ShcRNAi). High-fat diet feeding of wild-type mice amplified surface P-selectin expression on platelets stimulated by the thrombin receptor agonist protease-activated receptor-4 (PAR4), and increased aggregation of platelets induced by thrombin. These exaggerated platelet responses induced by high-fat diet feeding were significantly blunted in p66ShcRNAi mice. Finally, thrombin-stimulated platelet reactive oxygen species were suppressed in p66ShcRNAi mice. Hypercholesterolemia stimulates p66Shc expression in platelets, promoting platelet oxidative stress, hyperreactivity and hyperaggregation via p66Shc.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%