Merck
  • Chemical communication between cardiac cells is disrupted by high glucose: implications for the diabetic heart.
Chemical communication between cardiac cells is disrupted by high glucose: implications for the diabetic heart.
Experimental cell research (2014-09-28)
Walmor C De Mello
ABSTRACT
The influence of high glucose solution on the chemical communication between cardiac cells was investigated in cell pairs isolated from the left ventricle of adult Wistar Kyoto rats. For this, Lucifer Yellow CH was dialyzed into one cell of the pair using the whole cell clamp technique, and the diffusion of dye in the dialyzed as well as in non-dialyzed cell, was followed by measuring the intensity of fluorescence in both cells as a function of time. The results indicated that: 1) high glucose solution (25 mM) disrupted chemical communication between cardiac cells; 2) the effect of high glucose solution was reduced by Bis-1 (10(-9)M) which is a PKC inhibitor, and by enalapril (10(-9)M); 3) intracellular dialysis of Ang II (100 nM) also caused dye uncoupling; 4) calculation of gap junction permeability (Pj) (cm/s) indicated a value of 3 ± 0.07 × 10(-5)cm/s; n=32; (6 animals) for the controls and 0.4 ± 0.86 × 10(-6)cm/s; n=35 (6 animals) (P<0.05) for cells incubated with high glucose solution for 24h; 5) measurements of Pj for cell pairs treated with hypertonic solution plus Bis-1 (10(-9)M) or enalapril maleate (10(-9)M) showed no significant change of Pj (P>0.05). high glucose (25 mM) disrupts chemical communication between cardiac cells-an effect highly dependent on PKC activation. The possible role of enhanced intracellular Ang II levels induced by high glucose on the disruption of chemical communication was discussed as well as the possible implications of these findings for the diabetic heart.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Enalapril Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Enalapril maleate, United States Pharmacopeia (USP) Reference Standard
Enalapril maleate, European Pharmacopoeia (EP) Reference Standard
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%