Merck
  • A liquid chromatography-tandem mass spectrometry-based method for the simultaneous determination of hydroxy sterols and bile acids.
A liquid chromatography-tandem mass spectrometry-based method for the simultaneous determination of hydroxy sterols and bile acids.
Journal of chromatography. A (2014-12-03)
Clara John, Philipp Werner, Anna Worthmann, Katrin Wegner, Klaus Tödter, Ludger Scheja, Sascha Rohn, Joerg Heeren, Markus Fischer
ABSTRACT
Recently, hydroxy sterols and bile acids have gained growing interest as they are important regulators of energy homoeostasis and inflammation. The high number of different hydroxy sterols and bile acid species requires powerful analytical tools to quantify these structurally and chemically similar analytes. Here, we introduce a liquid chromatography-tandem mass spectrometry (LC-MS/MS)-based method for rapid quantification of 34 sterols (hydroxy sterols, primary, secondary bile acids as well as their taurine and glycine conjugates). Chromatographic baseline separation of isomeric hydroxy sterols and bile acids is obtained using a rugged amide embedded C18 (polar embedded) stationary phase. The current method features a simple extraction protocol validated for blood plasma, urine, gall bladder, liver, feces, and adipose tissue avoiding solid phase extraction as well as derivatization procedures. The total extraction recovery for representative analytes ranged between 58-86% in plasma, 85% in urine, 79-92% in liver, 76-98% in adipose tissue, 93-104% in feces and 62-79% in gall bladder. The validation procedure demonstrated that the calibration curves were linear over the selected concentration ranges for 97% of the analytes, with calculated coefficients of determination (R2) of greater than 0.99. A feeding study in wild type mice with a standard chow and a cholesterol-enriched Western type diet illustrated that the protocol described here provides a powerful tool to simultaneously quantify cholesterol derivatives and bile acids in metabolically active tissues and to follow the enterohepatic circulation.
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Stigmasterol, ~95%
Sigma-Aldrich
Desmosterol, ≥84% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
5β-Pregnan-3α-ol-20-one
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Progesterone, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Glycoursodeoxycholic acid, ≥96.0% (TLC)
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)