Merck
  • Development of a novel inhalational model of invasive pulmonary aspergillosis in rats and comparative evaluation of three biomarkers for its diagnosis.
Development of a novel inhalational model of invasive pulmonary aspergillosis in rats and comparative evaluation of three biomarkers for its diagnosis.
PloS one (2014-06-24)
Suhail Ahmad, Ali A Al-Shaikh, Ziauddin Khan
ABSTRACT
Aspergillus fumigatus, a thermotolerant fungus, is the main causative agent of invasive pulmonary aspergillosis (IPA) in immunocompromised patients that is associated with high mortality rates. Early diagnosis of IPA is crucial for mortality reduction and improved prognosis. An experimental inhalational model of IPA was developed in rats and the efficacy of three biomarkers, namely β-D-glucan (BDG), a panfungal marker, galactomannan (GM), a genus-specific marker, and A. fumigatus DNA, a species-specific marker was evaluated in serum and bronchoalveolar lavage (BAL) specimens at different time points postinfection for early diagnosis of IPA. BDG and GM were detected by using commercial Fungitell and Platelia Aspergillus EIA kits, respectively. A. fumigatus DNA was detected by developing a sensitive, single-step PCR assay. IPA was successfully developed in immunosuppressed rats and all animals until 5 days post-infection were positive for A. fumigatus by culture and KOH-calcofluor microscopy also showed A. fumigatus in 19 of 24 (79%) lung tissue samples. Fourteen of 30 (47%) and 27 of 30 (90%) serum and BAL specimens, respectively, were positive for all three biomarkers with 100% specificity (none of sera or BAL specimens of 12 control rats was positive for biomarkers). Our data show that BAL is a superior specimen than serum and combined detection of BDG, GM and A. fumigatus DNA provide a sensitive diagnosis of IPA in an experimental animal model. Moreover, combined detection of GM and DNA in BAL and detection of either GM or DNA in serum was also positive in 27 of 30 (90%) animals. For economic reasons and considering that the positive predictive value of BDG is low, the detection of GM and/or DNA in serum and BAL samples has the potential to serve as an integral component of the diagnostic-driven strategy in high-risk patients suspected for IPA.
Product Description
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C