Merck
  • Hydrogen peroxide reactions on cocaine in hair using imaging mass spectrometry.
Hydrogen peroxide reactions on cocaine in hair using imaging mass spectrometry.
Forensic science international (2014-07-23)
Eva Cuypers, Bryn Flinders, Ingrid J Bosman, Klaas J Lusthof, Arian C Van Asten, Jan Tytgat, Ron M A Heeren
ABSTRACT
Today, forensic hair analysis is considered to be a standard method for identifying chronic drug users since information about drug use stored and located in hair can cover several months to even years. When interpreting these results, one should be aware of all kind of pitfalls. External factors such as bleaching might influence the analytical result. Although the effect of hydrogen peroxide on cocaine in a solution was described before, it was never investigated whether the described reaction products (ecgonine methylester, benzoylecgonine, hydroxynorcocaine and dihydroxycocaine) are indeed found on contaminated or user hair. Since it is of great importance in forensic hair analysis to know whether cocaine and/or reaction products are detectable in hair after bleaching, matrix-assisted laser desorption/ionization mass spectrometric imaging (MALDI-MSI) was used to study the effect of hydrogen peroxide treatment on incorporated cocaine in hairs. Cocaine oxidation products were identified in a solution based on MS/MS spectra and spatial distribution of these products in hair was explored using MALDI TOF-MS. All images were accomplished by spraying α-Cyano-4-hydroxycinnamic acid (CHCA) as a MALDI-matrix. Images revealed a loss of detectability of cocaine and its reaction products in hairs already after a short bleaching period. Since all compounds of interest are found in the hydrogen peroxide and wash solution, these findings indicate that all evidence of cocaine use might be lost after a hair bleaching treatment. Therefore, forensic toxicologists should take into consideration whether hair samples were bleached before making any conclusions from hair analysis results.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard