Merck
  • Comprehensive two-dimensional liquid chromatography coupled to high resolution time of flight mass spectrometry for chemical characterization of sewage treatment plant effluents.
Comprehensive two-dimensional liquid chromatography coupled to high resolution time of flight mass spectrometry for chemical characterization of sewage treatment plant effluents.
Journal of chromatography. A (2015-01-13)
Xiyu Ouyang, Pim Leonards, Juliette Legler, Ron van der Oost, Jacob de Boer, Marja Lamoree
ABSTRACT
For the first time a comprehensive two-dimensional liquid chromatography (LC×LC) system coupled with a high resolution time-of-flight mass spectrometer (HR-ToF MS) was developed and applied for analysis of emerging toxicants in wastewater effluent. The system was optimized and validated using environmental standard compound mixtures of e.g. carbamate pesticides and polycyclic aromatic hydrocarbons (PAHs), to characterize the chromatographic system, to test the stability of the retention times and orthogonality. Various stationary phases in the second dimension were compared for the LC×LC analysis of silicon rubber passive sampler extracts of a wastewater effluent. A combination of C18 and Pentafluorophenyl (PFP) was found to be most effective. Finally, the hyphenation of LC×LC with HR-ToF MS was optimized, including splitter settings, transfer of data files between the different software packages and background subtraction using instrument software tools, after which tentative identification of 20 environmental contaminants was achieved, including pesticides, pharmaceuticals and food additives. As examples, three pesticides (isoproturon, terbutryn and diazinon) were confirmed by two-dimensional retention alignment.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%