Merck
  • Pathologic and molecular profiling of rapid-onset fibrosis and inflammation induced by multi-walled carbon nanotubes.
Pathologic and molecular profiling of rapid-onset fibrosis and inflammation induced by multi-walled carbon nanotubes.
Archives of toxicology (2014-12-17)
Jie Dong, Dale W Porter, Lori A Batteli, Michael G Wolfarth, Diana L Richardson, Qiang Ma
ABSTRACT
Multi-walled carbon nanotubes (MWCNT) are new materials with a wide range of industrial and commercial applications. However, their nano-scaled size and fiber-like shape render them respirable and potentially fibrogenic if inhaled into the lungs. To understand MWCNT fibrogenesis, we analyzed the pathologic and molecular aspects of the early phase response to MWCNT in mouse lungs. MWCNT induced rapid and pronounced lesions in the lungs characterized by increased cellularity and formation of fibrotic foci, most notably near where MWCNT deposited, within 14 days post-exposure. Deposition of collagen fibers was markedly increased in the alveolar septa and fibrotic foci, accompanied by elevated expression of fibrotic genes Col1a1, Col1a2, and Fn1 at both mRNA and protein levels. Fibrosis was induced rapidly at 40 μg, wherein fibrotic changes were detected on day 1 and reached a maximal intensity on day 7 through day 14. Induction of fibrosis was dose-dependent at the dose range of 5-40 μg, 7 days post-exposure. MWCNT elicited rapid and prominent infiltrations of neutrophils and macrophages alongside fibrosis implicating acute inflammation in the fibrotic response. At the molecular level, MWCNT induced elevated expression of proinflammatory cytokines TNFα, IL1α, IL1β, IL6, and CCL2 in lung tissues as well as the bronchoalveolar lavage fluid, in a dose- and time-dependent manner. MWCNT also increased the expression of fibrogenic growth factors TGF-β1 and PDGF-A in the lungs significantly. These findings underscore the interplay between acute inflammation and the early fibrotic response in the initiation and propagation of pulmonary fibrosis induced by MWCNT.
Product Description
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%