Merck
  • Overexpression of microRNA-125b sensitizes human hepatocellular carcinoma cells to 5-fluorouracil through inhibition of glycolysis by targeting hexokinase II.
Overexpression of microRNA-125b sensitizes human hepatocellular carcinoma cells to 5-fluorouracil through inhibition of glycolysis by targeting hexokinase II.
Molecular medicine reports (2014-05-29)
Jian-Xin Jiang, Shan Gao, Yao-Zhen Pan, Chao Yu, Cheng-Yi Sun
ABSTRACT
5-fluorouracil (5-FU)-based chemotherapy is widely used in the treatment of human hepatocellular carcinoma. However, despite impressive initial clinical responses, the majority of patients eventually develop resistance to 5-FU. The microRNA (miR)-125 family has been implicated in a variety of carcinomas as either a tumor suppressor or promoter. In the present study, the role of miR-125b in acquired 5-FU resistance in multiple human hepatocellular carcinoma cell lines was investigated using transfection of miR-125b. Compared with 5-FU?sensitive cells, 5?FU?resistant cells exhibited reduced expression levels of miR?125b. Furthermore, transfection of pre?miR-125b into liver cancer cells resulted in sensitization of 5-FU?resistant cells to 5-FU. In addition, the glucose uptake and lactate production in 5-FU?resistant liver cancer cells were demonstrated to be significantly increased compared with 5?FU?sensitive cells (P<0.05), indicating that targeting glycolytic pathways may overcome chemoresistance in human liver cancer cells. Notably, miR-125 was found to downregulate glucose metabolism by directly targeting hexokinase II. Since drug resistance is a common phenotype of malignant cancer cells, the finding that miR-125b expression levels are negatively correlated with 5-FU resistance in hepatocellular carcinoma cells is consistent with the reported functions of miR-125b. In conclusion, the present study identified miR-125b as a tumor suppressor-like microRNA, which exhibits great potential as a diagnostic and prognostic biomarker in hepatocellular carcinoma.
Sigma-Aldrich
O-Phosphorylethanolamine
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Williams′ Medium E, With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture