Merck
  • Functional nucleus pulposus-like matrix assembly by human mesenchymal stromal cells is directed by macromer concentration in photocrosslinked carboxymethylcellulose hydrogels.
Functional nucleus pulposus-like matrix assembly by human mesenchymal stromal cells is directed by macromer concentration in photocrosslinked carboxymethylcellulose hydrogels.
Cell and tissue research (2014-08-06)
Michelle S Gupta, Steven B Nicoll
ABSTRACT
Intervertebral disc (IVD) degeneration is associated with several pathophysiologic changes of the IVD, including dehydration of the nucleus pulposus (NP). Tissue engineering strategies may be used to restore both biological and mechanical function of the IVD following removal of NP tissue during surgical intervention. Recently, photocrosslinked carboxymethylcellulose (CMC) hydrogels were shown to support chondrogenic, NP-like extracellular matrix (ECM) elaboration by human mesenchymal stromal cells (hMSCs) when supplemented with TGF-β3; however, mechanical properties of these constructs did not reach native values. Fabrication parameters (i.e., composition, crosslinking density) can influence the bulk mechanical properties of hydrogel scaffolds, as well as cellular behavior and differentiation patterns. The objective of this study was to evaluate the influence of CMC macromer concentration (1.5, 2.5 and 3.5 % weight/volume) on bulk hydrogel properties and NP-like matrix elaboration by hMSCs. The lowest macromer concentration of 1.5 % exhibited the highest gene expression levels of aggrecan and collagen II at day 7, corresponding with the largest accumulation of glycosaminoglycans and collagen II by day 42. The ECM elaboration in the 1.5 % constructs was more homogeneously distributed compared to primarily pericellular localization in 3.5 % gels. The 1.5 % gels also displayed significant improvements in mechanical functionality by day 42 compared to earlier time points, which was not seen in the other groups. The effects of macromer concentration on matrix accumulation and organization are likely attributed to quantifiable differences in polymer crosslinking density and diffusive properties between the various hydrogel formulations. Taken together, these results demonstrate that macromer concentration of CMC hydrogels can direct hMSC matrix elaboration, such that a lower polymer concentration allows for greater NP-like ECM assembly and improvement of mechanical properties over time.
Product Description
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
1,9-Dimethyl-Methylene Blue zinc chloride double salt, Dye content 80 %
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
SAFC
L-Proline
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis