Merck
  • Identification of effective combinatorial markers for quality standardization of herbal medicines.
Identification of effective combinatorial markers for quality standardization of herbal medicines.
Journal of chromatography. A (2014-05-06)
Zi-Qi Shi, De-Fang Song, Ruo-Qian Li, Hua Yang, Lian-Wen Qi, Gui-Zhong Xin, Ding-Qian Wang, Hui-Peng Song, Jun Chen, Haiping Hao, Ping Li
ABSTRACT
Quality standardization of herbal medicines (HMs) is an important task with great challenges. Selection of abundant compounds as markers is currently a major approach for the quality control of HMs; however, such marker compounds are irrelevant to the bioactivities in many cases. Taking Lycoridis Radiatae Bulbus (LRB) as an example, we proposed a universal strategy to identify the effective combinatorial markers (ECMs) that are representative of the bioactivities of HMs, and took them as chemical markers for quality standardization. Fingerprinting and quantification were employed to find out the common components in various batches of medicines. The contribution of each common compound to the overall bioactivity was determined through fingerprint-bioactivity modeling, which based on the absolute quantification of each compound and the acetylcholinesterase (AChE) inhibitory activity of LRB. Two most effective compounds, ungerimine and galanthamine, were therefore proposed as ECMs. Interestingly, these two compounds could synergistically inhibit AChE. This approach demonstrated its strong advantage of the bioactivity relevant quality assessment when compared with conventional methods. And the success of applying this ECMs-based method to the quality assessment of unknown LRB samples proved that our approach was reliable and reproducible. In conclusion, this approach is not only useful for the bioactivity relevant quality control of HMs but also helpful for the discovery of ECMs as new drug candidates.
Product Description
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups