Merck
  • Synthetic (+)-terrein suppresses interleukin-6/soluble interleukin-6 receptor induced-secretion of vascular endothelial growth factor in human gingival fibroblasts.
Synthetic (+)-terrein suppresses interleukin-6/soluble interleukin-6 receptor induced-secretion of vascular endothelial growth factor in human gingival fibroblasts.
Bioorganic & medicinal chemistry (2014-08-26)
Hiroki Mandai, Kazuhiro Omori, Daisuke Yamamoto, Toki Tsumura, Kyouta Murota, Satoshi Yamamoto, Koichi Mitsudo, Soichiro Ibaragi, Akira Sasaki, Hiroshi Maeda, Shogo Takashiba, Seiji Suga
ABSTRACT
Interleukin (IL)-6 is a proinflammatory cytokine that performs a wide variety of biological functions, including important roles in the progression of chronic inflammatory diseases such as periodontal disease. (+)-Terrein, a secondary bioactive fungal metabolite isolated from Aspergillus terreus, has various biological activities; however, its anti-inflammatory effects are still unknown. The purpose of this study was to examine the effect of synthetic (+)-terrein on IL-6 signaling and related protein production in human gingival fibroblasts. To our knowledge, this study is the first to report that synthetic (+)-terrein is not cytotoxic at concentrations less than 20 μM and suppresses IL-6/soluble IL-6 receptor (sIL-6R)-induced phosphorylation of signal transducer and activator of transcription-3, extracellular signal-regulated kinase 1/2, and c-jun N-terminal kinase 1/2-signaling proteins that are downstream of IL-6 signaling. In addition, synthetic (+)-terrein suppresses IL-6/sIL-6R-induced vascular endothelial growth factor (VEGF) secretion in a concentration-dependent manner (p<0.01). These data suggest that synthetic (+)-terrein has potential anti-IL-6 signaling activity and suppresses VEGF-associated inflammatory disease progression.
Product Description
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Rhein
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
tert-Butylchlorodimethylsilane solution, 50 wt. % in toluene
Sigma-Aldrich
Tetrabutylammonium fluoride on silica gel, ~1.5 mmol/g (F-)
Sigma-Aldrich
Dimethyl methylphosphonate, ≥98%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Dimethyl methylphosphonate, purum, ≥97.0% (GC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material