Merck
  • Regulation and function of deiodinases during decidualization in female mice.
Regulation and function of deiodinases during decidualization in female mice.
Endocrinology (2014-05-07)
Wen-Bo Deng, Xiao-Huan Liang, Ji-Long Liu, Zeng-Ming Yang
ABSTRACT
Thyroid dysfunction during human pregnancy is closely related to serious pregnancy outcome. However, the regulation and function of thyroid hormones during early pregnancy are largely unknown. We found that type II deiodinase, an enzyme converting T4 to activated T3, is highly expressed in the mouse uterus on days 3 and 4 of pregnancy. Once the embryo implants into the receptive uterus, type III deiodinase (Dio3), a mainly paternally imprinted gene for inactivating T3, is significantly induced in the stromal cells and accompanied by DNA hypermethylation of intergenic differentially CpG methylation regions in the δ-like 1 homolog-Dio3 imprinting cluster. The concentration of uterine free T3 is actually decreased after embryo implantation. T3 induces Dio3 expression both in vivo and in vitro, suggesting a positive feedback loop. T3 addition or Dio3 knockdown compromises decidualization. These results indicate that the Dio3-mediated local T3 decrease is critical for decidualization of stromal cells during early pregnancy. Furthermore, we found that progesterone regulates Dio3 expression through its cognate receptor both in vivo and in vitro. Additionally, cAMP regulates Dio3 transcription through the protein kinase A-cAMP response element-binding protein pathway. The inhibition of the protein kinase A pathway results in decreased Dio3 expression and impaired decidualization. Dio3 opposite strand (Dio3os) expressed in a similar pattern to Dio3, is transcribed from the opposite strand of Dio3 and fine-tunes Dio3 expression during decidualization. Our data indicate that Dio3 is strongly expressed and tightly controlled during decidualization.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dio3
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
USP
Sodium metabisulfite, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Supelco
Sodium metabisulfite, analytical standard
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%
Supelco
Sodium Metabisulfite, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
MISSION® esiRNA, targeting human DIO3
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydroquinone, meets USP testing specifications