Merck
Isolation and transplantation of sturgeon early-stage germ cells.
Theriogenology (2015-01-07)
Martin Pšenička, Taiju Saito, Zuzana Linhartová, Ievgeniia Gazo
ABSTRACT
We report, for the first time, a series of baseline techniques comprising isolation and transplantation of female and male early-stage germ cells in sturgeon to generate a germline chimera as a potential tool for surrogate reproduction and gene banking. Cells were dissociated from testis, characterized by mostly spermatogonia, and from ovary, exclusively comprising oogonia and previtellogenic oocytes, of Acipenser baerii, using 0.3% trypsin (2 hours, 23 °C) dissolved in PBS, isotonic with blood plasma. The dissociated germ cells were sorted by Percoll gradient centrifugation followed by immunolabeling with germ cell-specific vasa antibody DDX4, while 10% to 30% Percoll solution contained 79.4% and 70.8% labeled testicular and ovarian cells. Sorted germ cells were transplanted into a cavity close to a presumptive genital ridge of newly hatched heterospecific Acipenser ruthenus larvae with fluorescein isothiocyanate-labeled endogenous primordial germ cells. The transplanted germ cells were randomly distributed in the body cavity through 30-day posttransplantation (dpt). Subsequently, the cells were organized into genital ridges 50 dpt and proliferated 90 dpt. The number of both transplanted and endogenous germ cells significantly increased from 18.1, 22.2, and 29.1 (30 dpt) to 108.5, 90.8, and 118.5 (90 dpt) in ovarian, testicular, and endogenous germ cells, respectively (P < 0.05). The efficiency of transplantation was 60% (counted 90 dpt).
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-15 Medium (Leibovitz), Without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sucrose, European Pharmacopoeia (EP) Reference Standard
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)