Merck
  • CTX-M-1 β-lactamase expression in Escherichia coli is dependent on cefotaxime concentration, growth phase and gene location.
CTX-M-1 β-lactamase expression in Escherichia coli is dependent on cefotaxime concentration, growth phase and gene location.
The Journal of antimicrobial chemotherapy (2014-09-04)
Thea S B Kjeldsen, Martin Overgaard, Søren S Nielsen, Valeria Bortolaia, Lotte Jelsbak, Morten Sommer, Luca Guardabassi, John E Olsen
ABSTRACT
Knowledge about the regulatory mechanisms of CTX-M β-lactamase-encoding genes in Escherichia coli is limited. The objectives of this study were to determine the growth response of CTX-M-1-producing E. coli exposed to cefotaxime and to investigate how blaCTX-M-1 expression at mRNA and protein levels is influenced by cefotaxime concentration, growth phase and gene location (chromosome versus plasmid). Two isogenic E. coli strains, MG1655/CTX-M-1 and MG1655/IncI1/CTX-M-1, containing blaCTX-M-1 on the chromosome and on a wild-type IncI1 plasmid, respectively, were constructed and the MIC of cefotaxime was determined. Growth of the two strains was studied in the presence of increasing concentrations of cefotaxime ranging from 0 to 512 mg/L. The levels of mRNA and protein in different growth phases and at different cefotaxime concentrations were studied by qPCR and selected-reaction-monitoring MS, respectively. The MICs of cefotaxime were 168 and 252 mg/L for MG1655/CTX-M-1 and MG1655/IncI1/CTX-M-1, respectively. Both strains displayed a prolonged lag phase when exposed to cefotaxime. The mRNA of blaCTX-M-1 and CTX-M-1 protein levels increased in the presence of high cefotaxime concentrations and varied with growth phase. Higher mRNA expression levels were detected for MG1655/CTX-M-1 compared with MG1655/IncI1/CTX-M-1, but a higher protein level was found for MG1655/IncI1/CTX-M-1 compared with MG1655/CTX-M-1, the latter corresponding well with the higher MIC for this strain. blaCTX-M-1 mRNA expression and CTX-M-1 protein levels were dependent on cefotaxime concentration, growth phase and gene location. These results provide insight into the expression of cephalosporin resistance in CTX-M-1-producing E. coli, improving our understanding of the relationship between antimicrobial therapy and the expression of resistance mechanisms.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
SAFC
Iodoacetamide
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%