Merck
Placental immune response to apple allergen in allergic mothers.
Journal of reproductive immunology (2014-07-23)
Martina Sandberg Abelius, Uta Enke, Frauke Varosi, Heike Hoyer, Ekkehard Schleussner, Maria C Jenmalm, Udo R Markert
ABSTRACT
The immunological milieu in the placenta may be crucial for priming the developing foetal immune system. Early imbalances may promote the establishment of immune-mediated diseases in later life, including allergies. The initial exposure to allergens seems to occur in utero, but little is known about allergen-induced placental cytokine and chemokine release. The release of several cytokines and chemokines from placenta tissue after exposure to mast cell degranulator compound 48/80 or apple allergen in placentas from allergic and healthy mothers was to be analysed. Four placentas from women with apple allergy and three controls were applied in a placental perfusion model with two separate cotyledons simultaneously perfused with and without apple allergen (Mal d 1). Two control placentas were perfused with compound 48/80. In outflow, histamine was quantified spectrophotofluorometrically, IL-2, IL-4, IL-6, IL-10, TNF and IFN-γ by a cytometric multiplex bead array and IL-13 and CXCL10, CXCL11, CCL17 and CCL22 with an in-house multiplex Luminex assay. Compound 48/80 induced a rapid release of histamine, CXCL10, CXCL11, CCL17 and CCL22, but not of the other factors. Apple allergen induced a time-dependent release of IL-6 and TNF, but not of histamine, in placentas of women with apple allergy compared with the unstimulated cotyledon. CCL17 levels were slightly increased after allergen stimulation in control placentas. Allergens can induce placental cytokines and chemokines distinctly in allergic and healthy mothers. These mediators may affect the prenatal development of the immune system and modify the risk of diseases related to immune disorders in childhood such as allergies.
Product Description
USP
Amoxicillin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Benzamidine hydrochloride, 99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Heptane, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Supelco
Histamine, analytical standard
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)