Merck
  • Neuroprotective activity and acetylcholinesterase inhibition of five Amaryllidaceae species: a comparative study.
Neuroprotective activity and acetylcholinesterase inhibition of five Amaryllidaceae species: a comparative study.
Life sciences (2014-12-23)
Natalie Cortes, Rafael Andrés Posada-Duque, Rafael Alvarez, Fernando Alzate, Strahil Berkov, Gloria Patricia Cardona-Gómez, Edison Osorio
ABSTRACT
Amaryllidaceae alkaloids exhibit a wide range of physiological effects, of which the acetylcholinesterase (AChE) inhibitory activity is the most relevant. However, scientific evidence related to their neuroprotective effectiveness against glutamate-induced toxicity has been lacking. Thus, the purpose of this study was to conduct a comparative study of the neuroprotective activity and the AChE inhibitory activity of species of Amaryllidaceae. The neuroprotective activity against glutamate-induced toxicity was measured in rat cortical neurons and the Ellman method was employed for the quantification of acetylcholinesterase inhibitory activity of alkaloidal extracts of five species of Amaryllidaceae (Crinum jagus, Crinum bulbispermum, Hippeastrum barbatum, Hippeastrum puniceum and Zephyranthes carinata). The alkaloid Amaryllidaceae patterns based on GC/MS analyses were also investigated. The results showed that the alkaloidal extract from C. jagus presented a high neuroprotective activity in both pre- and post-treatments against a glutamate excitotoxic stimulus. Furthermore, the alkaloid extracts from C. jagus and Z. carinata revealed an inhibitory activity of AChE from the electric eel with IC50 values of 18.28±0.29 and 17.96±1.22μg/mL, respectively. In addition, 46 alkaloids were detected by GC/MS, and 20 of them were identified based on their mass spectra and retention index. The results suggest that the neuroprotective effects might be associated with lycorine and crinine-type alkaloids, whereas the acetylcholinesterase enzyme inhibitory activity could be related to galanthamine and lycorine-type alkaloids, although not based on synergistic processes. In summary, Amaryllidaceae species are sources of alkaloids with potential use for Alzheimer's disease.
Product Description
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Supelco
Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Codeine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%