Merck
  • Asenapine maleate in situ forming biodegradable implant: an approach to enhance bioavailability.
Asenapine maleate in situ forming biodegradable implant: an approach to enhance bioavailability.
International journal of pharmaceutics (2014-10-12)
Amelia M Avachat, Sayali S Kapure
ABSTRACT
Biodegradable injectable in-situ forming implants (ISFI) correspond to an alternative parenteral depot system to microspheres and surgical implants. Objective of present work was to formulate and evaluate long acting implant of asenapine maleate (ASM) using PLGA which would release drug uniformly for 21 days. PLGA 50:50 with different drug: polymer ratios were tried. N-methyl-2-pyrrolidone and dimethyl sulphoxide were used as organic solvents. The influence of various parameters viz. polymer concentration, solvent ratio, viscosity and morphology on formation of implant was investigated. In-vitro dissolution studies indicated that drug: polymer ratio of 1:2 and N-methyl-2-pyrrolidone (0.3ml) gave desired release profile, total cumulative drug released being 97.66% at the end of 21 days. Mathematical models point towards erosion mechanism with zero order kinetics. Ex-vivo studies confirmed the formation of implant in extensor digitorum muscle with desired drug release profile. In-vivo study was performed in Sprague- Dawley rats. Compared to marketed sublingual formulation area under curve of ASM implant was found to increase 2.215 fold. The Cmax was found to be 11ng/ml. Thus long acting ISFI of ASM was successfully formulated showing improved therapeutic results for the treatment of schizophrenia and bipolar disorders which could be a potentialsubstitute to marketed sublingual tablets.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Asenapine maleate, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous