Merck
  • Uptake of aristolochic acid I into Caco-2 cells by monocarboxylic acid transporters.
Uptake of aristolochic acid I into Caco-2 cells by monocarboxylic acid transporters.
Biological & pharmaceutical bulletin (2014-09-02)
Osamu Kimura, Koichi Haraguchi, Chiho Ohta, Nobuyuki Koga, Yoshihisa Kato, Tetsuya Endo
ABSTRACT
The uptake mechanism of aristolochic acid I (AAI) was investigated using Caco-2 cells cultured on dishes and permeable membranes. The uptake of AAI from the apical membrane of Caco-2 cells cultured on a dish was rapid, and a decrease in the pH of the incubation medium significantly increased uptake. Incubation at low temperature (4°C) and treatment with sodium azide (a metabolic inhibitor) or carbonylcyanide p-trifluoromethoxyphenylhydrazone (a protonophore) significantly inhibited the AAI uptake. Coincubation with L-lactic acid or benzoic acid, typical substrates for the proton-linked monocarboxylic acid transporters (MCTs), significantly decreased the AAI uptake, as did coincubation with α-cyano-4-hydroxycinnamate (an inhibitor of MCTs). Dixon plotting revealed the competitive inhibition of benzoic acid on the AAI uptake. To confirm the AAI uptake via MCTs, the apical-to-basolateral transport of AAI was investigated using the Caco-2 cells cultured on the permeable membranes. The transport of AAI at pH 6.0 was markedly higher than that at pH 7.4, and was significantly decreased by coincubation with benzoic acid. These results suggest that the uptake of AAI from the apical membrane of Caco-2 cells is mediated mainly by MCTs along with benzoic acid.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
SAFC
L-Proline
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
p-Aminohippuric acid, ≥99%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
Phthalic acid, analytical standard